siRNA Details
Gene Symbol: Rtn4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022982.2 | NP_075358.1 | 2 | 507 | ||
GenBank mRNA | AF283462.1 | - | 2 | 509 | ||
BC052317.1 | - | 2 | 505 | 6397765 | ||
BC058381.1 | - | 2 | 564 | 6829861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
