siRNA Details
Gene Symbol: Arl6ip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022989.4 | NP_075365.3 | 1, 2 | 433 | ||
GenBank mRNA | AK011070.1 | - | 1, 2 | 387 | ||
AK016267.1 | - | 1, 2 | 408 | |||
AK032740.1 | - | 1, 2 | 407 | |||
AK035521.1 | - | 1, 2 | 430 | |||
AK052010.1 | - | 1, 2 | 407 | |||
AK082824.1 | - | 1, 2 | 405 | |||
AK159990.1 | - | 1, 2 | 395 | |||
BC019550.1 | - | 1, 2 | 400 | 4210432 | ||
BC157980.1 | - | 1, 2 | 424 | 9088139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
