siRNA Details
Gene Symbol: Lrp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022993.3 | NP_075369.2 | 6, 7 | 1977 | ||
GenBank mRNA | AB042200.1 | - | 6, 7 | 1879 | ||
AK132169.1 | - | 1961 | ||||
AK144035.1 | - | 6, 7 | 1990 | |||
AK170325.1 | - | 6, 7 | 1951 | |||
BC011058.1 | - | 2, 3 | 257 | |||
BC052378.1 | - | 6, 7 | 1953 | 5696275 | ||
BC054447.1 | - | 6, 7 | 1945 | 6477698 | ||
EF437372.1 | - | 1, 2 | 86 |
