siRNA Details
Gene Symbol: Bean1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141922.1 | NP_001135394.1 | 4 | 721 | ||
NM_001141924.1 | NP_001135396.1 | 3 | 615 | |||
NM_001141925.1 | NP_001135397.1 | 2 | 506 | |||
GenBank mRNA | AF240460.1 | - | 1 | 12 | ||
AK134633.1 | - | 3 | 615 | |||
BC030426.1 | - | 2 | 584 | |||
BC138184.1 | - | 4 | 650 | 8861203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
