siRNA Details
Gene Symbol: Prrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022999.1 | NP_075375.1 | 2 | 194 | ||
GenBank mRNA | AK005453.1 | - | 3 | 238 | ||
BC025098.1 | - | 2 | 194 | 4159863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
