siRNA Details
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023045.2 | NP_075532.1 | 27 | 3241 | ||
GenBank mRNA | AJ297360.1 | - | 27 | 3147 | ||
AK028303.1 | - | 25 | 2986 | |||
AK032768.1 | - | 9 | 1116 | |||
AK035775.1 | - | 27 | 3166 | |||
AK036837.1 | - | 25 | 2986 | |||
AK122361.1 | - | 26 | 3184 | |||
AK143729.1 | - | 27 | 3212 | |||
AK164595.1 | - | 27 | 3169 | |||
BC029702.1 | - | 27 | 3167 | 5053250 |

More Information
Other Species Targeted

Gene Ontology Categories:
