siRNA Details
Gene Symbol: Asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039126.2 | NP_001034215.1 | 2 | 337 | ||
NM_023046.4 | NP_075533.3 | 2 | 138 | |||
GenBank mRNA | AF155352.1 | - | 2 | 89 | ||
AK006515.1 | - | 2 | 139 | |||
AK122456.1 | - | 2 | 193 | |||
AK153774.1 | - | 2 | 359 | |||
BC034869.1 | - | 2 | 193 | |||
BC049699.1 | - | 2 | 138 | |||
BU938650.1 | - | 2 | 141 | |||
BY231875.1 | - | 2 | 335 | |||
KC789642.1 | - | 2 | 170 | |||
KC789643.1 | - | 2 | 170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
