siRNA Details
Gene Symbol: Utp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023054.1 | NP_075541.1 | 1 | 1016 | ||
GenBank mRNA | AF155362.2 | - | 1 | 1011 | ||
AF271213.1 | - | 1 | 1016 | |||
AK010316.1 | - | 1 | 1033 | |||
AK145637.1 | - | 1 | 1029 | |||
BC003213.1 | - | 1 | 1033 | |||
BC016581.1 | - | 1 | 1034 | |||
BC022994.1 | - | 1 | 1021 | 5371012 |

More Information
Other Species Targeted

Gene Ontology Categories:
