siRNA Details
Gene Symbol: Utp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023054.1 | NP_075541.1 | 1 | 921 | ||
GenBank mRNA | AF155362.2 | - | 1 | 916 | ||
AF271213.1 | - | 1 | 921 | |||
AK010316.1 | - | 1 | 938 | |||
AK145637.1 | - | 1 | 934 | |||
BC003213.1 | - | 1 | 938 | |||
BC016581.1 | - | 1 | 939 | |||
BC022994.1 | - | 1 | 926 | 5371012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
