siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 2 | 466 | ||
NM_001164208.1 | NP_001157680.1 | 2 | 114 | |||
NM_001164209.2 | NP_001157681.1 | 3 | 322 | |||
NM_001286651.1 | NP_001273580.1 | 3 | 319 | |||
NM_001286652.1 | NP_001273581.1 | 2 | 226 | |||
NM_023056.4 | NP_075543.1 | 2 | 715 | |||
GenBank mRNA | AB031386.1 | - | 2 | 275 | ||
AF115426.1 | - | 2 | 187 | |||
AK007408.1 | - | 2 | 121 | |||
AK156042.1 | - | 2 | 467 | |||
AK177944.1 | - | 1 | 20 | |||
BC019539.1 | - | 1 | 84 | 4237639 | ||
BC022166.1 | - | 2 | 730 | 3582123 | ||
BQ571258.1 | - | 2 | 106 | |||
CT010176.1 | - | 1 | 79 | |||
CX213755.1 | - | 2 | 226 | |||
CX223561.1 | - | 3 | 302 | |||
DV047496.1 | - | 2 | 154 | |||
DV059241.1 | - | 2 | 190 |
