siRNA Details
Gene Symbol: Lima1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113545.1 | NP_001107017.1 | 7 | 1035 | ||
NM_023063.4 | NP_075550.2 | 4 | 907 | |||
GenBank mRNA | AF307844.1 | - | 4 | 830 | ||
AF307845.1 | - | 7 | 989 | |||
AK028186.1 | - | 4 | 848 | |||
AK031698.1 | - | 4 | 851 | |||
AK049350.1 | - | 7 | 985 | |||
AK085065.1 | - | 4 | 901 | |||
AK132314.1 | - | 4 | 853 | |||
AK167220.1 | - | 4 | 467 | |||
BC031490.1 | - | 7 | 961 | 3498138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
