siRNA Details
Gene Symbol: Tmem254b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270495.1 | NP_001257424.1 | 3, 4 | 520 | ||
NM_001270496.1 | NP_001257425.1 | 3, 4 | 278 | |||
GenBank mRNA | BY713360.1 | - | 3, 4 | 520 | ||
CK793021.1 | - | 3, 4 | 276 | |||
DV046977.1 | - | 2, 3 | 138 |
Gene Symbol: Tmem254a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025311.3 | NP_079587.1 | 367 | |||
NM_026679.2 | NP_080955.1 | 536 | ||||
GenBank mRNA | AK002342.1 | - | 3, 4 | 355 | ||
AK002441.1 | - | 3, 4 | 467 | |||
AK003554.1 | - | 3, 4 | 276 | |||
AK008299.1 | - | 3, 4 | 327 | |||
BC031854.1 | - | 3, 4 | 420 | 4194400 | ||
BY270140.1 | - | 368 |
Gene Symbol: Tmem254c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270498.1 | NP_001257427.1 | 536 | |||
NM_001270499.1 | NP_001257428.1 | 356 | ||||
GenBank mRNA | BY703810.1 | - | 276 | |||
CK128852.1 | - | 451 | ||||
CX201482.1 | - | 356 |
