siRNA Details
Gene Symbol: Dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025314.3 | NP_079590.1 | 4 | 500 | ||
GenBank mRNA | AK002246.1 | - | 4 | 436 | ||
AK003594.1 | - | 4 | 472 | |||
AK010822.1 | - | 4 | 438 | |||
AK011917.1 | - | 4 | 468 | |||
AK017565.1 | - | 4 | 424 | |||
AK158898.1 | - | 4 | 557 | |||
BC026537.1 | - | 4 | 473 | 5008303 |

More Information
Other Species Targeted

Gene Ontology Categories:
