siRNA Details
Gene Symbol: Sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025321.3 | NP_079597.2 | 5 | 311 | ||
GenBank mRNA | AK002459.1 | - | 5 | 313 | ||
AK012818.1 | - | 5 | 312 | |||
AK032458.1 | - | 313 | ||||
AK135503.1 | - | 5 | 313 | |||
BC083091.1 | - | 5 | 304 | 30445339 | ||
BY711932.1 | - | 5 | 312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
