siRNA Details
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 5 | 708 | ||
NM_023149.3 | NP_075638.2 | 6 | 676 | |||
GenBank mRNA | AB046738.1 | - | 6 | 617 | ||
AK003779.1 | - | 6 | 597 | |||
AK131124.1 | - | 5 | 588 | |||
AK149751.1 | - | 5 | 633 | |||
AK151013.1 | - | 6 | 598 | |||
AK152442.1 | - | 5 | 674 | |||
AK152729.1 | - | 5 | 646 | |||
AK153040.1 | - | 5 | 647 | |||
AK159129.1 | - | 5 | 675 | |||
AK159739.1 | - | 6 | 604 | |||
AK168029.1 | - | 6 | 615 | |||
AK168652.1 | - | 6 | 627 | |||
AK168856.1 | - | 4 | 365 | |||
AK172034.1 | - | 6 | 598 | |||
AK189897.1 | - | 3 | 245 | |||
BC005532.1 | - | 6 | 607 | 3496319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
