siRNA Details
Gene Symbol: Tmem176a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098271.1 | NP_001091741.1 | 5 | 611 | ||
NM_025326.4 | NP_079602.4 | 5 | 663 | |||
GenBank mRNA | AB063313.1 | - | 4 | 553 | ||
AK002546.1 | - | 5 | 639 | |||
AK079378.1 | - | 625 | ||||
AK150576.1 | - | 5 | 628 | |||
AK162991.1 | - | 3 | 1871 | |||
AK165660.1 | - | 5 | 637 | |||
AK180847.1 | - | 1 | 149 | |||
BC010831.1 | - | 5 | 607 | 4221626 | ||
CT010190.1 | - | 4 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
