siRNA Details
Gene Symbol: Tmem176a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098271.1 | NP_001091741.1 | 5 | 597 | ||
NM_025326.4 | NP_079602.4 | 5 | 649 | |||
GenBank mRNA | AB063313.1 | - | 4 | 539 | ||
AK002546.1 | - | 5 | 625 | |||
AK079378.1 | - | 611 | ||||
AK150576.1 | - | 5 | 614 | |||
AK162991.1 | - | 3 | 1857 | |||
AK165660.1 | - | 5 | 623 | |||
AK180847.1 | - | 1 | 135 | |||
BC010831.1 | - | 5 | 593 | 4221626 | ||
CT010190.1 | - | 4 | 539 |
