siRNA Details
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025327.2 | NP_079603.1 | 5 | 397 | ||
GenBank mRNA | AK002479.1 | - | 5 | 398 | ||
AK003310.1 | - | 5 | 365 | |||
BC031492.1 | - | 5 | 350 | 3485092 | ||
BC043030.1 | - | 5 | 869 | 5693076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
