siRNA Details
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023154.3 | NP_075643.1 | 3 | 672 | ||
GenBank mRNA | AB049623.1 | - | 2 | 125 | ||
AK002666.1 | - | 2 | 127 | |||
BC010592.1 | - | 2 | 130 | 3156338 | ||
BC083162.1 | - | 2 | 106 | 6529658 | ||
BC094044.1 | - | 3 | 672 | 5064931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
