siRNA Details
Gene Symbol: Tmem167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025335.3 | NP_079611.2 | 4 | 1245 | ||
GenBank mRNA | AK002884.1 | - | 4 | 1171 | ||
AK012161.1 | - | 4 | 1179 | |||
AK031706.1 | - | 4 | 1180 | |||
AK032474.1 | - | 4 | 1189 | |||
AK088895.1 | - | 4 | 1167 | |||
AK132361.1 | - | 4 | 1167 | |||
AK161037.1 | - | 4 | 1182 | |||
BC085489.1 | - | 4 | 1152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
