siRNA Details
Gene Symbol: Eif3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025344.2 | NP_079620.2 | 5 | 754 | ||
GenBank mRNA | AK002778.1 | - | 5 | 753 | ||
AK003372.1 | - | 5 | 752 | |||
AK010333.1 | - | 5 | 747 | |||
AK088537.1 | - | 5 | 754 | |||
AK136797.1 | - | 753 | ||||
AK152037.1 | - | 5 | 755 | |||
AK186458.1 | - | 3 | 240 | |||
AK190701.1 | - | 5 | 400 | |||
AK192917.1 | - | 4 | 348 | |||
AK200417.1 | - | 3 | 240 | |||
AK209599.1 | - | 3 | 238 | |||
AK214434.1 | - | 3 | 240 | |||
AK217928.1 | - | 4 | 372 | |||
AY061808.1 | - | 2 | 98 | |||
BC070473.1 | - | 5 | 736 | 30634857 | ||
BC083190.1 | - | 5 | 739 | 30358339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
