siRNA Details
Gene Symbol: Cxcl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023158.6 | NP_075647.3 | 4 | 987 | ||
GenBank mRNA | AF277001.1 | - | 4 | 450 | ||
AF301017.1 | - | 4 | 516 | |||
AK003085.1 | - | 4 | 646 | |||
AK009599.1 | - | 4 | 536 | |||
AK028875.1 | - | 4 | 989 | |||
AK030515.1 | - | 4 | 412 | |||
AK150224.1 | - | 4 | 675 | |||
AK151330.1 | - | 4 | 761 | |||
AK152102.1 | - | 4 | 761 | |||
AK152547.1 | - | 4 | 939 | |||
AK153192.1 | - | 4 | 703 | |||
AK170428.1 | - | 5 | 1158 | |||
GQ380016.1 | - | 5 | 553 | |||
GQ380017.1 | - | 6 | 604 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
