siRNA Details
Gene Symbol: Tceal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025355.4 | NP_079631.2 | 3 | 1070 | ||
GenBank mRNA | AK005297.1 | - | 3 | 1072 | ||
BC087890.1 | - | 3 | 1070 | 6533845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
