siRNA Details
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025356.4 | NP_079632.1 | 6, 7 | 546 | ||
GenBank mRNA | AK003163.1 | - | 6, 7 | 497 | ||
AK004001.1 | - | 6, 7 | 556 | |||
AK009276.1 | - | 6, 7 | 488 | |||
AK009936.1 | - | 6, 7 | 485 | |||
AK032885.1 | - | 6, 7 | 556 | |||
AK049906.1 | - | 6, 7 | 547 | |||
AK077666.1 | - | 6, 7 | 606 | |||
AK088448.1 | - | 6, 7 | 516 | |||
AK151295.1 | - | 6, 7 | 660 | |||
AK151594.1 | - | 6, 7 | 567 | |||
AK152231.1 | - | 6, 7 | 567 | |||
AK152541.1 | - | 6, 7 | 540 | |||
AK153100.1 | - | 6, 7 | 660 | |||
AK153111.1 | - | 6, 7 | 568 | |||
AK153410.1 | - | 6, 7 | 567 | |||
AK153423.1 | - | 6, 7 | 606 | |||
AK159125.1 | - | 6, 7 | 606 | |||
AK159546.1 | - | 6, 7 | 542 | |||
AK159963.1 | - | 6, 7 | 540 | |||
AK165235.1 | - | 5, 6 | 787 | |||
AK167077.1 | - | 6, 7 | 556 | |||
AK167138.1 | - | 6, 7 | 509 | |||
AK167650.1 | - | 6, 7 | 544 | |||
AK167801.1 | - | 6, 7 | 568 | |||
AK169177.1 | - | 5, 6 | 918 | |||
BC023266.1 | - | 5, 6 | 311 | |||
BC024351.1 | - | 7, 8 | 595 | |||
BC057941.1 | - | 6, 7 | 527 | 5323636 | ||
BC098220.1 | - | 6, 7 | 561 | 963757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
