siRNA Details
Gene Symbol: Marc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290273.1 | NP_001277202.1 | 4 | 888 | ||
GenBank mRNA | AK004989.1 | - | 4 | 785 | ||
AK011722.1 | - | 4 | 769 | |||
BC028441.1 | - | 2 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
