siRNA Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025364.2 | NP_079640.1 | 8, 9 | 598 | ||
GenBank mRNA | AK003453.1 | - | 8, 9 | 450 | ||
AK018773.1 | - | 8, 9 | 461 | |||
AK088266.1 | - | 8, 9 | 447 | |||
AK146248.1 | - | 8, 9 | 476 | |||
BC027510.1 | - | 8, 9 | 437 | 1382354 | ||
BC157909.1 | - | 8, 9 | 462 | 9088111 | ||
BC158023.1 | - | 8, 9 | 462 | 9007329 | ||
BC172063.1 | - | 8, 9 | 459 | 9054349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
