siRNA Details
Gene Symbol: Tomm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164729.1 | NP_001158201.1 | 3 | 868 | ||
NM_025365.3 | NP_079641.1 | 3 | 814 | |||
GenBank mRNA | AK003291.1 | - | 3 | 476 | ||
AK010470.1 | - | 3 | 526 | |||
AK019199.1 | - | 3 | 476 | |||
BC021590.1 | - | 2 | 1000 | 5068609 | ||
BC037589.1 | - | 3 | 515 | 5007624 | ||
BC089325.1 | - | 1 | 1662 | |||
CA319049.1 | - | 3 | 517 | |||
CA321392.1 | - | 3 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
