siRNA Details
Gene Symbol: Wbscr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025375.3 | NP_079651.2 | 3, 4 | 337 | ||
GenBank mRNA | AF412035.1 | - | 2, 3 | 109 | ||
AK003386.1 | - | 2, 3 | 110 | |||
AK011005.1 | - | 2, 3 | 117 | |||
AK151986.1 | - | 2, 3 | 118 | |||
BC093484.1 | - | 2, 3 | 87 | 6735279 | ||
BC117986.1 | - | 3, 4 | 337 | 40102067 | ||
BC117987.1 | - | 3, 4 | 337 | 40102068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
