siRNA Details
Gene Symbol: Atp6v1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025381.2 | NP_079657.1 | 2 | 227 | ||
GenBank mRNA | AK003419.1 | - | 2 | 224 | ||
AK007327.1 | - | 2 | 228 | |||
AK152153.1 | - | 2 | 224 | |||
AK152929.1 | - | 2 | 251 | |||
AK152959.1 | - | 2 | 224 | |||
BC016553.1 | - | 2 | 237 | 2812592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
