siRNA Details
Gene Symbol: Dnajc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025384.3 | NP_079660.1 | 4 | 302 | ||
GenBank mRNA | AK002598.1 | - | 4 | 298 | ||
AK003396.1 | - | 4 | 312 | |||
AK007806.1 | - | 4 | 291 | |||
AK007833.1 | - | 4 | 291 | |||
BC027509.1 | - | 4 | 302 | 1364626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
