siRNA Details
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025387.3 | NP_079663.1 | 6 | 604 | ||
GenBank mRNA | AK003891.1 | - | 6 | 553 | ||
AK008495.1 | - | 5 | 494 | |||
AK008878.1 | - | 6 | 526 | |||
AK010609.1 | - | 6 | 564 | |||
AK012101.1 | - | 6 | 551 | |||
AK019395.1 | - | 6 | 553 | |||
AK050688.1 | - | 6 | 539 | |||
AK211061.1 | - | 2 | 198 | |||
AK219176.1 | - | 2 | 198 | |||
BC025854.1 | - | 6 | 505 | 5250659 | ||
BC055862.1 | - | 6 | 528 | 3492090 | ||
BC069899.1 | - | 5 | 1250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
