siRNA Details
Gene Symbol: Nip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164472.1 | NP_001157944.1 | 1, 2 | 116 | ||
NM_025391.2 | NP_079667.2 | 1, 2 | 116 | |||
GenBank mRNA | AK003739.1 | - | 1, 2 | 116 | ||
AK006057.1 | - | 1, 2 | 95 | |||
AK014290.1 | - | 1, 2 | 75 | |||
BC003972.1 | - | 1, 2 | 103 | 3488726 | ||
BE623972.1 | - | 1, 2 | 87 |
