siRNA Details
Gene Symbol: Tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183250.2 | NP_899073.1 | 3 | 157 | ||
GenBank mRNA | AK002863.1 | - | 3 | 158 | ||
AK003762.1 | - | 3 | 158 | |||
AK160705.1 | - | 3 | 155 | |||
BC034602.1 | - | 3 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
