siRNA Details
Gene Symbol: Mustn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181390.3 | NP_852055.1 | 3 | 549 | ||
GenBank mRNA | AJ277212.1 | - | 3 | 685 | ||
AK003360.1 | - | 3 | 265 | |||
AK003945.1 | - | 3 | 267 | |||
BC116939.1 | - | 3 | 257 | 40126258 | ||
BC116941.1 | - | 3 | 257 | 40126260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
