siRNA Details
Gene Symbol: Nop10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025403.4 | NP_079679.1 | 2 | 234 | ||
GenBank mRNA | AK004120.1 | - | 2 | 235 | ||
AK012875.1 | - | 2 | 191 | |||
BC028497.1 | - | 2 | 189 | 1398559 |

More Information
Other Species Targeted

Gene Ontology Categories:
