siRNA Details
Gene Symbol: Sptssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164210.2 | NP_001157682.1 | 4 | 264 | ||
NM_001286959.1 | NP_001273888.1 | 3 | 233 | |||
NM_133675.3 | NP_598436.1 | 3 | 233 | |||
GenBank mRNA | AF365876.1 | - | 3 | 235 | ||
AK004017.1 | - | 3 | 235 | |||
AK009973.1 | - | 4 | 231 | |||
AK010117.1 | - | 3 | 234 | |||
BC022628.1 | - | 3 | 213 | 4206004 | ||
CX729774.1 | - | 3 | 207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
