siRNA Details
Gene Symbol: 1110037F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081183.1 | NP_001074652.1 | 5 | 459 | ||
GenBank mRNA | AK034232.1 | - | 5 | 381 | ||
AK052499.1 | - | 5 | 397 | |||
AK076731.1 | - | 5 | 429 | |||
AK122509.1 | - | 5 | 362 | |||
AK155623.1 | - | 5 | 460 | |||
AK162531.1 | - | 5 | 428 | |||
AK170075.1 | - | 5 | 462 | |||
BC033309.1 | - | 5 | 429 | 5024976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
