siRNA Details
Gene Symbol: 1110037F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081183.1 | NP_001074652.1 | 18, 19 | 4471 | ||
GenBank mRNA | AK122509.1 | - | 17, 18 | 4224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
