siRNA Details
Gene Symbol: Ier3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025409.3 | NP_079685.1 | 2 | 266 | ||
GenBank mRNA | AK004281.1 | - | 2 | 251 | ||
AK007388.1 | - | 2 | 214 | |||
AK007862.1 | - | 2 | 198 | |||
AK009116.1 | - | 2 | 199 | |||
AK190140.1 | - | 2 | 137 | |||
AK210106.1 | - | 2 | 134 | |||
BC021512.1 | - | 2 | 256 | 5351700 | ||
CX207811.1 | - | 2 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
