siRNA Details
Gene Symbol: Them5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025416.3 | NP_079692.1 | 3 | 510 | ||
GenBank mRNA | AK003517.1 | - | 2 | 77 | ||
AK004155.1 | - | 3 | 511 | |||
AK009228.1 | - | 3 | 509 | |||
BC046437.1 | - | 3 | 492 | 3969280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
