siRNA Details
Gene Symbol: Commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033132.3 | NP_001028304.1 | 3 | 225 | ||
NM_001168592.1 | NP_001162064.1 | 2 | 213 | |||
GenBank mRNA | AK004316.1 | - | 3 | 199 | ||
AK008220.1 | - | 3 | 207 | |||
AK009796.1 | - | 3 | 220 | |||
AK011226.1 | - | 3 | 210 | |||
AK018711.1 | - | 3 | 199 | |||
AK019351.1 | - | 3 | 201 | |||
AK019432.1 | - | 3 | 202 | |||
AK020234.1 | - | 3 | 198 | |||
AK034223.1 | - | 1 | 3473 | |||
AK045407.1 | - | 2699 | ||||
BC132261.1 | - | 3 | 212 | 40130538 | ||
BC132265.1 | - | 3 | 212 | 40130542 | ||
BC145360.1 | - | 3 | 212 | 9053918 | ||
BC145361.1 | - | 2 | 200 | 9053919 | ||
CK621880.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
