siRNA Details
Gene Symbol: 1110059G10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025419.4 | NP_079695.2 | 2 | 304 | ||
GenBank mRNA | AK004312.1 | - | 2 | 300 | ||
AK012115.1 | - | 2 | 300 | |||
AK083316.1 | - | 2 | 277 | |||
AK160617.1 | - | 2 | 305 | |||
BC034133.1 | - | 2 | 262 | 4921846 |
