siRNA Details
Gene Symbol: 1110059G10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025419.4 | NP_079695.2 | 3 | 475 | ||
GenBank mRNA | AK004312.1 | - | 3 | 471 | ||
AK083316.1 | - | 3 | 448 | |||
AK160617.1 | - | 3 | 476 | |||
AK220157.1 | - | 2 | 283 | |||
BC034133.1 | - | 3 | 433 | 4921846 |
