siRNA Details
Gene Symbol: 1110059E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025423.2 | NP_079699.1 | 3, 4 | 399 | ||
GenBank mRNA | AK004308.1 | - | 3, 4 | 400 | ||
AK016266.1 | - | 3, 4 | 546 | |||
BC023385.1 | - | 3, 4 | 396 | 5253886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
