siRNA Details
Gene Symbol: 1110059E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025423.2 | NP_079699.1 | 2 | 253 | ||
GenBank mRNA | AK004308.1 | - | 2 | 254 | ||
AK016266.1 | - | 2 | 400 | |||
BC023385.1 | - | 2 | 250 | 5253886 |
