siRNA Details
Gene Symbol: Nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025424.2 | NP_079700.1 | 2, 3 | 265 | ||
GenBank mRNA | AB126218.1 | - | 2, 3 | 239 | ||
AK002413.1 | - | 2, 3 | 266 | |||
AK004329.1 | - | 2, 3 | 254 | |||
AK009304.1 | - | 2, 3 | 256 | |||
AK020403.1 | - | 2, 3 | 247 | |||
AK155319.1 | - | 2, 3 | 264 | |||
BC048464.1 | - | 2, 3 | 253 | 6596336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
