siRNA Details
Gene Symbol: Inip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013577.1 | NP_001013595.1 | 3 | 272 | ||
GenBank mRNA | AK004256.1 | - | 2 | 145 | ||
AK005245.1 | - | 3 | 254 | |||
AK012023.1 | - | 3 | 192 | |||
AK159116.1 | - | 3 | 260 | |||
AK169537.1 | - | 2 | 174 | |||
BB641174.1 | - | 3 | 272 | |||
BC116204.1 | - | 3 | 167 | 40090152 | ||
BC116205.1 | - | 3 | 167 | 40090155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
