siRNA Details
Gene Symbol: Med7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104530.1 | NP_001098000.1 | 3 | 455 | ||
NM_001104556.1 | NP_001098026.1 | 2 | 453 | |||
NM_001104557.1 | NP_001098027.1 | 2 | 285 | |||
NM_025426.3 | NP_079702.3 | 3 | 623 | |||
GenBank mRNA | AK004354.1 | - | 3 | 588 | ||
AK010357.1 | - | 3 | 451 | |||
AK012799.1 | - | 3 | 603 | |||
AK144918.1 | - | 2 | 411 | |||
AK147739.1 | - | 3 | 613 | |||
AK167407.1 | - | 3 | 583 | |||
BC011333.1 | - | 3 | 610 | 3993084 | ||
BE333513.1 | - | 2 | 383 | |||
C88719.1 | - | 2 | 258 | |||
CK030901.1 | - | 3 | 455 | |||
CK788351.1 | - | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
