siRNA Details
Gene Symbol: Zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037762.1 | NP_001032851.1 | 5 | 694 | ||
NM_025428.2 | NP_079704.2 | 5 | 736 | |||
GenBank mRNA | AK004475.1 | - | 5 | 637 | ||
AK013074.1 | - | 5 | 724 | |||
AK087676.1 | - | 5 | 708 | |||
BC021432.1 | - | 5 | 676 | 5030263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
