siRNA Details
Gene Symbol: Trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025432.3 | NP_079708.2 | 4 | 410 | ||
GenBank mRNA | AK002658.1 | - | 5 | 481 | ||
AK004389.1 | - | 435 | ||||
AK007889.1 | - | 4 | 411 | |||
AK007951.1 | - | 345 | ||||
AK008564.1 | - | 5 | 460 | |||
AK077643.1 | - | 436 | ||||
AY318751.1 | - | 468 | ||||
BC034845.1 | - | 445 | 1528500 | |||
BC061087.1 | - | 480 | 6589013 | |||
BC099551.1 | - | 452 | 30252145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
